[(2-Chlorethoxy)methyl]benzol 98%

Lieferant: Apollo Scientific

Synonyme: 2-(Benzyloxy)ethyl Chloride, ((2-Chloroethoxy)methyl)benzene, (2-Chloroethoxymethyl)benzene

OR937793-25G
APOSOR937793-25GEA 59.6 EUR
APOSOR937793-25G
[(2-Chlorethoxy)methyl]benzol 98%
[(2-Chlorethoxy)methyl]benzol
Formel: C₉H₁₁ClO
Molecular Weight: 170.64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00018964
CAS-Nummer: 17229-17-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR