4-Nitrophenylacetat 95%
Lieferant: Apollo Scientific
OR937811-100G
APOSOR937811-100GEA
105
EUR
APOSOR937811-100G
4-Nitrophenylacetat 95%
4-Nitrophenylacetat
Formel:
C₈H₇NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007326 CAS-Nummer: 830-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...