3-Acryloyl-2-oxazolidinone 98%
Lieferant: Apollo Scientific
Synonyme:
3-Acryloyloxazolidin-2-one
OR937850-1G
APOSOR937850-1GEA
410
EUR
APOSOR937850-1G
3-Acryloyl-2-oxazolidinone 98%
3-Acryloyl-2-oxazolidinone
Formel:
C₆H₇NO₃ Molecular Weight: 141,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038407 CAS-Nummer: 2043-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...