[2-(2-Methoxyphenoxy)ethyl]methylamine 97%

Lieferant: Apollo Scientific

OR937878-1G
APOSOR937878-1GEA 290 EUR
APOSOR937878-1G
[2-(2-Methoxyphenoxy)ethyl]methylamine 97%
[2-(2-Methoxyphenoxy)ethyl]methylamine
Formel: C₁₀H₁₅NO₂
Molecular Weight: 181,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07400597
CAS-Nummer: 72955-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR