4-Penten-1-yl isothiocyanate 95%
Lieferant: Apollo Scientific
OR937905-1G
OR937905-250MG
APOSOR937905-1GEA
319
EUR
APOSOR937905-1G
APOSOR937905-250MG
4-Penten-1-yl isothiocyanate 95%
4-Penten-1-yl isothiocyanate
Formel:
C₆H₉NS Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191527 CAS-Nummer: 18060-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...