4-Penten-1-yl isothiocyanate 95%

Lieferant: Apollo Scientific

OR937905-1G OR937905-250MG
APOSOR937905-1GEA 319 EUR
APOSOR937905-1G APOSOR937905-250MG
4-Penten-1-yl isothiocyanate 95%
4-Penten-1-yl isothiocyanate
Formel: C₆H₉NS
Lagertemperatur: Raumtemperatur
MDL: MFCD00191527
CAS-Nummer: 18060-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR