1,2-Dihydroxyindan 95%

Lieferant: Apollo Scientific

OR937916-10G OR937916-1G OR937916-5G
APOSOR937916-10GEA 683 EUR
APOSOR937916-10G APOSOR937916-1G APOSOR937916-5G
1,2-Dihydroxyindan 95%
1,2-Dihydroxyindan
Formel: C₉H₁₀O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 4370-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR