Ethylene glycol mono-m-tolyl ether 98%
Lieferant: Apollo Scientific
OR937919-5G
APOSOR937919-5GEA
0
EUR
APOSOR937919-5G
Ethylene glycol mono-m-tolyl ether 98%
Ethylene glycol mono-m-tolyl ether
Formel:
C₉H₁₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014035 CAS-Nummer: 13605-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...