Allylpalladium(II) chloride dimer 97%

Lieferant: Apollo Scientific
Achtung

OR937920-1G
APOSOR937920-1GEA 0 EUR
APOSOR937920-1G
Allylpalladium(II) chloride dimer 97%
Allylpalladium(II) chloride dimer
Formel: C₆H₁₀Cl₂Pd₂
Lagertemperatur: Kühlschrank
MDL: MFCD00044874
CAS-Nummer: 12012-95-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR