2-Chloro-N-(R)-(1-phenylethyl)acetamide 95%

Lieferant: Apollo Scientific

OR937941-1G
APOSOR937941-1GEA 342 EUR
APOSOR937941-1G
2-Chloro-N-(R)-(1-phenylethyl)acetamide 95%
2-Chloro-N-(R)-(1-phenylethyl)acetamide
Formel: C₁₀H₁₂ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00040691
CAS-Nummer: 36293-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR