Diethylene glycol monododecyl ether 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Dodecyl Carbitol, 2-(2-Dodecyloxyethoxy)ethanol

OR938002-1G
APOSOR938002-1GEA 693 EUR
APOSOR938002-1G
Diethylene glycol monododecyl ether 95%
Diethylene glycol monododecyl ether
Formel: C₁₆H₃₄O₃
Molecular Weight: 274,45 g/mol
Schmelzpunkt: 16 °C
MDL: MFCD00042658
CAS-Nummer: 3055-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR