1,6-Naphthyridin-2(1H)-one 97%

Lieferant: Apollo Scientific

OR938005-1G OR938005-250MG
APOSOR938005-1GEA 632 EUR
APOSOR938005-1G APOSOR938005-250MG
1,6-Naphthyridin-2(1H)-one 97%
1,6-Naphthyridin-2(1H)-one
Formel: C₈H₆N₂O
Molecular Weight: 146.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781179
CAS-Nummer: 23616-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR