2-Cyclopentylethanol 95%

Lieferant: Apollo Scientific

OR938017-5G
APOSOR938017-5GEA 30.6 EUR
APOSOR938017-5G
2-Cyclopentylethanol 95%
2-Cyclopentylethanol
Formel: C₇H₁₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00019301
CAS-Nummer: 766-00-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR