1,5-Dimethyl-2-piperidinon 95%

Lieferant: Apollo Scientific

OR938034-10G OR938034-1G OR938034-5G
APOSOR938034-10GEA 754 EUR
APOSOR938034-10G APOSOR938034-1G APOSOR938034-5G
1,5-Dimethyl-2-piperidinon 95%
1,5-Dimethyl-2-piperidone
Formel: C₇H₁₃NO
Molecular Weight: 127.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701098
CAS-Nummer: 86917-58-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR