N-Methylacetanilid 98%
Lieferant: Apollo Scientific
OR938036-25G
APOSOR938036-25GEA
63.8
EUR
APOSOR938036-25G
N-Methylacetanilid 98%
N-Methylacetanilid
Formel:
C₉H₁₁NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00026181 CAS-Nummer: 579-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...