cis-4-Hydroxycyclohexancarbonsäure 98%

Lieferant: Apollo Scientific

OR938037-25G
APOSOR938037-25GEA 190 EUR
APOSOR938037-25G
cis-4-Hydroxycyclohexancarbonsäure 98%
cis-4-Hydroxycyclohexanecarboxylic acid
Formel: C₇H₁₂O₃
Molecular Weight: 144.1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093477
CAS-Nummer: 3685-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR