3,3,5,5-Tetramethyl-1-pyrroline-N-oxide 95%

Lieferant: Apollo Scientific

OR938091-1G
APOSOR938091-1GEA 683 EUR
APOSOR938091-1G
3,3,5,5-Tetramethyl-1-pyrroline-N-oxide 95%
3,3,5,5-Tetramethyl-1-pyrroline-N-oxide
Formel: C₈H₁₅NO
Molecular Weight: 141,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010542
CAS-Nummer: 10135-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR