Potassium isopropylxanthate 95%

Lieferant: Apollo Scientific

OR938096-25G OR938096-5G
APOSOR938096-25GEA 98.6 EUR
APOSOR938096-25G APOSOR938096-5G
Potassium isopropylxanthate 95%
Potassium isopropylxanthate
Formel: C₄H₈OS₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00004930
CAS-Nummer: 140-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR