4-Isopropylthiazole-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR938097-1G
APOSOR938097-1GEA 567 EUR
APOSOR938097-1G
4-Isopropylthiazole-2-carboxylic acid 95%
4-Isopropylthiazole-2-carboxylic acid
Formel: C₇H₉NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09864275
CAS-Nummer: 300831-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR