Methyl-(3,4-dihydroxyphenyl)acetat 98%

Lieferant: Apollo Scientific

Synonyme: 3,4-Dihydroxyphenylacetic Acid Methyl Ester, Methyl 2-(3,4-dihydroxyphenyl)acetate, Methyl 3,4-dihydroxyphenylacetate, Catechol-4-acetic Acid Methyl Ester, Homoprotocatechuic Acid Methyl Ester, Pyrocatechol-4-acetic Acid Methyl Ester

OR938109-5G
APOSOR938109-5GEA 95.7 EUR
APOSOR938109-5G
Methyl-(3,4-dihydroxyphenyl)acetat 98%
Methyl-(3,4-dihydroxyphenyl)acetat
Formel: C₉H₁₀O₄
Molecular Weight: 182.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00583539
CAS-Nummer: 25379-88-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR