N-(n-Propyl)-2-(2-pyridyl)ethylamine 97%
Lieferant: Apollo Scientific
OR938116-1G
APOSOR938116-1GEA
522
EUR
APOSOR938116-1G
N-(n-Propyl)-2-(2-pyridyl)ethylamine 97%
N-(n-Propyl)-2-(2-pyridyl)ethylamine
Formel:
C₁₀H₁₆N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023544 CAS-Nummer: 55496-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...