3-Chlor-2,2-dimethylpropansäure 97%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-2,2-dimethylpropionic Acid, 3-Chloro-2,2-dimethylpropanoic acid

OR938167-25G
APOSOR938167-25GEA 177 EUR
APOSOR938167-25G
3-Chlor-2,2-dimethylpropansäure 97%
3-Chlor-2,2-dimethylpropansäure
Formel: C₅H₉ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00004195
CAS-Nummer: 13511-38-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR