3-Anilino-1-propanol 95%

Lieferant: Apollo Scientific

Synonyme: N-(3-Hydroxypropyl)aniline, 3-(Phenylamino)propan-1-ol

OR938169-5G
APOSOR938169-5GEA 208 EUR
APOSOR938169-5G
3-Anilino-1-propanol 95%
3-Anilino-1-propanol
Formel: C₉H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD15206074
CAS-Nummer: 31121-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR