2-Naphthylacetat 95%

Lieferant: Apollo Scientific

Synonyme: Essigsäure(2-naphthyl)ester, β-Naphthylacetat, Essigsäure(β-naphthyl)ester

OR938172-25G
APOSOR938172-25GEA 69.6 EUR
APOSOR938172-25G
2-Naphthylacetat 95%
2-Naphthylacetat
Formel: C₁₂H₁₀O₂
Molecular Weight: 186,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004060
CAS-Nummer: 1523-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR