4-Methoxy-3-(piperidin-1-ylsulfonyl)phenylboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Methoxy-3-(piperidin-1-ylsulphonyl)phenyl)boronic acid

OR9382-1G
APOSOR9382-1GEA 263 EUR
APOSOR9382-1G
4-Methoxy-3-(piperidin-1-ylsulfonyl)phenylboronsäure 95%
4-Methoxy-3-(piperidin-1-ylsulfonyl)phenylboronsäure
Formel: C₁₂H₁₈BNO₅S
Molecular Weight: 299,1500 g/mol
Schmelzpunkt: 157…159 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235037
CAS-Nummer: 874219-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR