Sodium-2-mercaptobenzothiazole 97%

Lieferant: Apollo Scientific
Achtung

OR938282-25G OR938282-100G OR938282-500G
APOSOR938282-25GEA 21.9 EUR
APOSOR938282-25G APOSOR938282-100G APOSOR938282-500G
Sodium-2-mercaptobenzothiazole 97%
Sodium-2-mercaptobenzothiazole
Formel: C₇H₅NS₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00065334
CAS-Nummer: 2492-26-4
UN: 3077
ADR: 9+EHS,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR