Sodium-2-mercaptobenzothiazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR938282-25G
OR938282-100G
OR938282-500G
APOSOR938282-25GEA
21.9
EUR
APOSOR938282-25G
APOSOR938282-100G
APOSOR938282-500G
Sodium-2-mercaptobenzothiazole 97%
Sodium-2-mercaptobenzothiazole
Formel:
C₇H₅NS₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065334 CAS-Nummer: 2492-26-4 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...