Pent-4-yn-1-amine hydrochloride 97%

Lieferant: Apollo Scientific

OR938293-500MG
APOSOR938293-500MGEA 346 EUR
APOSOR938293-500MG
Pent-4-yn-1-amine hydrochloride 97%
Pent-4-yn-1-amine hydrochloride
Formel: C₅H₉N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD22836507
CAS-Nummer: 173987-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR