2-(Dimethylcarbamoyl)phenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Dimethylcarbamoyl)benzeneboronic acid
OR9383-1G
APOSOR9383-1GEA
278
EUR
APOSOR9383-1G
2-(Dimethylcarbamoyl)phenylboronsäure 95%
2-(Dimethylcarbamoyl)phenylboronsäure
Formel:
C₉H₁₂BNO₃ Molecular Weight: 193,01 g/mol Schmelzpunkt: 108…110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03425959 CAS-Nummer: 874219-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...