1-(Ammoniooxy)propanchlorid 95%

Lieferant: Apollo Scientific

OR938341-1G OR938341-250MG
APOSOR938341-1GEA 770 EUR
APOSOR938341-1G APOSOR938341-250MG
1-(Ammoniooxy)propanchlorid 95%
1-(Ammoniooxy)propanchlorid
Formel: C₃H₁₀ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00231456
CAS-Nummer: 6084-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR