1,2,3,4-Tetrahydroisoquinolin-8-ol 95%

Lieferant: Apollo Scientific

OR938375-1G OR938375-250MG
APOSOR938375-1GEA 638 EUR
APOSOR938375-1G APOSOR938375-250MG
1,2,3,4-Tetrahydroisoquinolin-8-ol 95%
1,2,3,4-Tetrahydroisoquinolin-8-ol
Formel: C₉H₁₁NO
Lagertemperatur: Kühlschrank
MDL: MFCD11506139
CAS-Nummer: 32999-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR