2-(4-Morpholinylcarbonyl)phenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR9384-1G
APOSOR9384-1GEA
321
EUR
APOSOR9384-1G
2-(4-Morpholinylcarbonyl)phenylboronsäure 95%
2-(4-Morpholinylcarbonyl)phenylboronsäure
Formel:
C₁₁H₁₄BNO₄ Schmelzpunkt: 150…152 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235036 CAS-Nummer: 874219-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...