3-Methoxyphthalid 95%
Lieferant: Apollo Scientific
Synonyme:
3-Methoxyisobenzofuran-1(3H)-one, 3-Methoxy-1(3H)-isobenzofuranone
OR938427-5G
APOSOR938427-5GEA
163
EUR
APOSOR938427-5G
3-Methoxyphthalid 95%
3-Methoxyphthalid
Formel:
C₉H₈O₃ Molecular Weight: 164,1580 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01652196 CAS-Nummer: 4122-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...