Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate 95%

Lieferant: Apollo Scientific

OR938432-1G
APOSOR938432-1GEA 72.5 EUR
APOSOR938432-1G
Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate 95%
Methyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate
Formel: C₁₁H₁₃NO₂
Molecular Weight: 191,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03981003
CAS-Nummer: 57060-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR