Ethyl-4-formylbenzoate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-formylbenzoate

OR938434-1G
APOSOR938434-1GEA 94.4 EUR
APOSOR938434-1G
Ethyl-4-formylbenzoate 95%
Ethyl-4-formylbenzoate
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00460463
CAS-Nummer: 6287-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR