Di(1-adamantyl)-n-butylphosphine hydriodide 97%

Lieferant: Apollo Scientific
Achtung

OR938437-1G OR938437-5G OR938437-25G
APOSOR938437-1GEA 66.7 EUR
APOSOR938437-1G APOSOR938437-5G APOSOR938437-25G
Di(1-adamantyl)-n-butylphosphine hydriodide 97%
Di(1-adamantyl)-n-butylphosphine hydriodide
Formel: C₂₄H₄₀IP
Lagertemperatur: Gefrierschrank
MDL: MFCD10566973
CAS-Nummer: 714951-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR