1,3-Dimethyl-1H-pyrazole-4-sulphonyl chloride 95%

Lieferant: Apollo Scientific

OR938453-5G
APOSOR938453-5GEA 256 EUR
APOSOR938453-5G
1,3-Dimethyl-1H-pyrazole-4-sulphonyl chloride 95%
1,3-Dimethyl-1H-pyrazole-4-sulphonyl chloride
Formel: C₅H₇ClN₂O₂S
Molecular Weight: 194.64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03944999
CAS-Nummer: 89501-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR