6-Chloro-n,n-dimethylnicotinamide 95%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-N,N-dimethylnicotinamide

OR938495-1G
APOSOR938495-1GEA 60.9 EUR
APOSOR938495-1G
6-Chloro-n,n-dimethylnicotinamide 95%
6-Chloro-n,n-dimethylnicotinamide
Formel: C₈H₉ClN₂O
Molecular Weight: 184,623 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08588286
CAS-Nummer: 54864-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR