5-Brom-N-(2-hydroxyethyl)-2-methoxybenzolsulfonamid 98%

Lieferant: Apollo Scientific

OR9385-1G
APOSOR9385-1GEA 168 EUR
APOSOR9385-1G
5-Brom-N-(2-hydroxyethyl)-2-methoxybenzolsulfonamid 98%
5-Brom-N-(2-hydroxyethyl)-2-methoxybenzolsulfonamid
Formel: C₉H₁₂BrNO₄S
Schmelzpunkt: 109…112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08056375
CAS-Nummer: 871269-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR