2-(Chlormethyl)-2-ethyloxiran 95%
Lieferant: Apollo Scientific
OR938507-1G
OR938507-250MG
OR938507-5G
APOSOR938507-1GEA
203
EUR
APOSOR938507-1G
APOSOR938507-250MG
APOSOR938507-5G
2-(Chlormethyl)-2-ethyloxiran 95%
2-(Chloromethyl)-1,2-epoxybutane
Formel:
C₅H₉ClO Lagertemperatur: Raumtemperatur |
CAS-Nummer:
75484-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...