2-(Chlormethyl)-2-ethyloxiran 95%

Lieferant: Apollo Scientific

OR938507-1G OR938507-250MG OR938507-5G
APOSOR938507-1GEA 203 EUR
APOSOR938507-1G APOSOR938507-250MG APOSOR938507-5G
2-(Chlormethyl)-2-ethyloxiran 95%
2-(Chloromethyl)-1,2-epoxybutane
Formel: C₅H₉ClO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 75484-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR