5-Isopropyl-2-pyrimidinamine 95%

Lieferant: Apollo Scientific

OR938512-1G
APOSOR938512-1GEA 1640 EUR
APOSOR938512-1G
5-Isopropyl-2-pyrimidinamine 95%
5-Isopropyl-2-pyrimidinamine
Formel: C₇H₁₁N₃
Molecular Weight: 137,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08460249
CAS-Nummer: 98432-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR