2-Methylbenzhydryl chloride 98%
Lieferant: Apollo Scientific
OR938518-5G
APOSOR938518-5GEA
59.6
EUR
APOSOR938518-5G
2-Methylbenzhydryl chloride 98%
2-Methylbenzhydryl chloride
Formel:
C₁₄H₁₃Cl Lagertemperatur: Kühlschrank |
MDL:
MFCD00972765 CAS-Nummer: 41870-52-4 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...