1-(1-Methyl-1H-pyrazol-4-yl)ethanone 95%
Lieferant: Apollo Scientific
Synonyme:
1-(1-Methyl-1h-pyrazol-4-yl)-ethanone
OR938532-5G
APOSOR938532-5GEA
60.9
EUR
APOSOR938532-5G
1-(1-Methyl-1H-pyrazol-4-yl)ethanone 95%
1-(1-Methyl-1H-pyrazol-4-yl)ethanone
Formel:
C₆H₈N₂O Molecular Weight: 124.141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159640 CAS-Nummer: 37687-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...