2-(Propylamino)ethanol 95%

Lieferant: Apollo Scientific

Synonyme: N-Propylethanolamine

OR938537-5G
APOSOR938537-5GEA 74.1 EUR
APOSOR938537-5G
2-(Propylamino)ethanol 95%
2-(Propylamino)ethanol
Formel: C₅H₁₃NO
Molecular Weight: 103.16 g/mol
Siedepunkt: 182
Dichte: 0.901
Lagertemperatur: Raumtemperatur
MDL: MFCD00002844
CAS-Nummer: 16369-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR