(S)-1-(3,4-Dimethoxyphenyl)ethylamine 95%

Lieferant: Apollo Scientific

Synonyme: (S)-1-(3,4-Dimethoxyphenyl)ethanamine

OR938552-1G
APOSOR938552-1GEA 104 EUR
APOSOR938552-1G
(S)-1-(3,4-Dimethoxyphenyl)ethylamine 95%
(S)-1-(3,4-Dimethoxyphenyl)ethylamine
Formel: C₁₀H₁₅NO₂
Molecular Weight: 181.232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06761896
CAS-Nummer: 65451-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR