2-(Dimethylamino)propan-1-ol 95%

Lieferant: Apollo Scientific

OR938625-1G
APOSOR938625-1GEA 224 EUR
APOSOR938625-1G
2-(Dimethylamino)propan-1-ol 95%
2-(Dimethylamino)propan-1-ol
Formel: C₅H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00014425
CAS-Nummer: 15521-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR