Ethyl 1-hydroxycyclohexanecarboxylate 95%
Lieferant: Apollo Scientific
OR938628-1G
APOSOR938628-1GEA
0
EUR
APOSOR938628-1G
Ethyl 1-hydroxycyclohexanecarboxylate 95%
Ethyl 1-hydroxycyclohexanecarboxylate
Formel:
C₉H₁₆O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797659 CAS-Nummer: 1127-01-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...