2-Methyl-6-phenylpyridin 95%

Lieferant: Apollo Scientific

OR938642-1G
APOSOR938642-1GEA 221 EUR
APOSOR938642-1G
2-Methyl-6-phenylpyridin 95%
2-Methyl-6-phenylpyridine
Formel: C₁₂H₁₁N
Molecular Weight: 169.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00796537
CAS-Nummer: 46181-30-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR