2,3-Thiophenedicarboxylic anhydride 95%

Lieferant: Apollo Scientific

OR938656-1G
APOSOR938656-1GEA 0 EUR
APOSOR938656-1G
2,3-Thiophenedicarboxylic anhydride 95%
2,3-Thiophenedicarboxylic anhydride
Formel: C₆H₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD09832453
CAS-Nummer: 6007-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR